Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105379547 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030121.1 | XP_016885610.1 | 6 | 1513 |
Gene Symbol: LOC105371191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023917.1 | XP_016879406.1 | 7 | 1715 |