Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQCF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143833.3 | NP_001137305.2 | 1 | 365 | ||
XM_005265168.3 | XP_005265225.1 | 2 | 932 | |||
XM_011533728.1 | XP_011532030.1 | 2 | 403 | |||
GenBank mRNA | BX093815.1 | - | 1 | 365 |