Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM26F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010919.2 | NP_001010919.1 | 3 | 882 | ||
NM_001276460.1 | NP_001263389.1 | 2 | 299 | |||
XM_005266999.3 | XP_005267056.1 | 2 | 1166 | |||
XM_011535845.2 | XP_011534147.1 | 2 | 1246 | |||
GenBank mRNA | AF086130.1 | - | 1 | 204 | ||
AV734646.2 | - | 2 | 299 | |||
BC146842.1 | - | 2 | 808 | 9056770 |