Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RP9P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003500.1 | 4 | 446 | |||
GenBank mRNA | AF274938.1 | - | 3 | 156 | ||
BC050570.1 | - | 5 | 551 |