Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AARD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025357.2 | NP_001020528.1 | 2 | 567 | ||
XM_011517043.2 | XP_011515345.1 | 2 | 945 | |||
GenBank mRNA | AB181519.1 | - | 2 | 548 | ||
BC140924.1 | - | 2 | 593 | 9056736 | ||
BC142621.1 | - | 2 | 567 | 40147448 |