Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC107986910 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017014151.1 | XP_016869640.1 | 3 | 1036 | ||
XM_017014153.1 | XP_016869642.1 | 3 | 530 |
Gene Symbol: LOC101928095 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017014145.1 | XP_016869634.1 | 3 | 2652 |
Gene Symbol: LOC101929400 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017014147.1 | XP_016869636.1 | 3 | 1092 |