Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD19P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026868.1 | 9 | 1751 | |||
GenBank mRNA | AK093497.1 | - | 7 | 1343 | ||
BC028712.1 | - | 9 | 1715 |