Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CISD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008388.4 | NP_001008389.1 | 3 | 465 | ||
GenBank mRNA | AK292134.1 | - | 3 | 402 | ||
BC032300.1 | - | 3 | 402 | 5105935 | ||
BX537971.1 | - | 3 | 403 | |||
DN993861.1 | - | 3 | 470 |