Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PABPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002568.3 | NP_002559.2 | 15 | 2811 | ||
XM_005250861.2 | XP_005250918.1 | 15 | 3739 | |||
GenBank mRNA | AL135187.1 | - | 1 | 374 | ||
BC015958.2 | - | 16 | 2706 | 3940309 | ||
BC041863.1 | - | 15 | 3433 | |||
Y00345.1 | - | 15 | 2799 |