Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PCDH11X SIRNA Assays ›
Gene Symbol:
PCDH11X
Gene Name:
protocadherin 11 X-linked
Gene Aliases:
PCDH-X, PCDH11, PCDHX, PPP1R119
Chromosome Location:
Chr.X: 91779310 - 92623230 on Build GRCh38
UniGene ID:
Hs.655673
Species:
Human
siRNA ID
HSS167105
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PCDH11X
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001168360.1
NP_001161832.1
2
2079
NM_001168361.1
NP_001161833.1
2
2079
NM_001168362.1
NP_001161834.1
2
2079
NM_001168363.1
NP_001161835.1
2
2079
NM_032968.4
NP_116750.1
2
2079
NM_032969.4
NP_116751.1
2
2079
XM_011530910.1
XP_011529212.1
5
2554
XM_011530911.2
XP_011529213.1
7
2842
XM_011530912.2
XP_011529214.1
6
2788
XM_011530913.2
XP_011529215.1
6
2788
XM_011530914.2
XP_011529216.1
3
2454
XM_011530915.2
XP_011529217.1
6
2788
XM_011530916.2
XP_011529218.1
5
2554
XM_017029416.1
XP_016884905.1
2
2063
XM_017029417.1
XP_016884906.1
6
2788
XM_017029418.1
XP_016884907.1
6
2788
XM_017029419.1
XP_016884908.1
2
2063
XM_017029420.1
XP_016884909.1
6
2788
XM_017029421.1
XP_016884910.1
2
2063
XM_017029422.1
XP_016884911.1
3
2454
GenBank mRNA
AB026187.1
-
2
2880
AF206516.1
-
2
2035
AF217288.1
-
2
2035
AF332218.1
-
2
2880
AF332219.1
-
2
2880
AJ276804.1
-
5
2624
AJ564933.1
-
1
1495
AJ564934.1
-
1
1495
AJ564935.1
-
1
1495
AJ564936.1
-
1
1495
AK299813.1
-
5
2554
AK316263.1
-
5
2554
AY861432.1
-
2
2035
AY861433.1
-
2
2035
AY861434.1
-
2
2035
AY861435.1
-
2
2035
U79247.1
-
1
1093
Back To Top
More Information
Panther Classification:
Molecular Function -
cadherin
Gene Ontology Categories:
Function(s)
Process(es)
homophilic cell adhesion via plasma membrane adhesion molecules
negative regulation of phosphatase activity
calcium ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0