Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3orf70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025266.1 | NP_001020437.1 | 1, 2 | 378 | ||
GenBank mRNA | BC137178.1 | - | 1, 2 | 245 | 9021175 | |
BC137179.1 | - | 1, 2 | 245 | 9021176 | ||
BC150565.1 | - | 1, 2 | 256 | 40127634 |