Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMS2P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130940.1 | 2 | 195 | |||
GenBank mRNA | AB017006.1 | - | 2 | 290 | ||
DB472687.1 | - | 2 | 195 |
Gene Symbol: LOC441259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC075816.1 | - | 2 | 134 |
Gene Symbol: PMS2P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB017005.1 | - | 3 | 332 |