Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2J4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003655.3 | 4 | 427 | |||
GenBank mRNA | AK296066.1 | - | 3 | 230 | ||
BC017250.2 | - | 3 | 1329 | |||
BC062722.1 | - | 4 | 386 | |||
BC105584.1 | - | 4 | 409 | |||
BX537878.1 | - | 4 | 382 | |||
BX538054.1 | - | 3 | 206 | |||
CD674494.1 | - | 2 | 124 | |||
DA147922.1 | - | 4 | 427 |
Gene Symbol: DTX2P1-UPK3BP1-PMS2P11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023383.1 | 10 | 1533 | |||
GenBank mRNA | U38980.1 | - | 10 | 1418 |
Gene Symbol: LOC441259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC075816.1 | - | 6 | 575 |
Gene Symbol: PMS2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003613.1 | 6 | 1011 | |||
GenBank mRNA | AB017004.1 | - | 6 | 1011 | ||
D38435.1 | - | 6 | 614 | |||
D38499.1 | - | 6 | 614 |
Gene Symbol: PMS2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA282042.1 | - | 2 | 233 |
Gene Symbol: PMS2P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028059.1 | 9 | 1405 | |||
GenBank mRNA | D38437.1 | - | 5 | 553 | ||
D38498.1 | - | 5 | 553 |
Gene Symbol: PMS2P4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_022007.2 | 5 | 503 | |||
NR_046297.1 | 5 | 503 | ||||
NR_046299.1 | 5 | 503 | ||||
GenBank mRNA | BC029368.1 | - | 5 | 480 | ||
CB141754.1 | - | 5 | 468 | |||
D38438.1 | - | 5 | 472 | |||
D38500.1 | - | 5 | 472 | |||
DA885243.1 | - | 5 | 504 |
Gene Symbol: PMS2P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027775.2 | 6 | 620 | |||
NR_027776.2 | 5 | 517 | ||||
GenBank mRNA | AA621085.1 | - | 2 | 242 | ||
BF984173.1 | - | 5 | 516 | |||
D38436.1 | - | 6 | 601 | |||
D38502.1 | - | 6 | 601 |
Gene Symbol: PMS2P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB017005.1 | - | 2 | 170 |
Gene Symbol: PMS2P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130940.1 | 6 | 648 | |||
GenBank mRNA | AB017006.1 | - | 6 | 741 |
Gene Symbol: PMS2P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028058.1 | 4 | 908 | |||
GenBank mRNA | BM845848.1 | - | 5 | 449 |
Gene Symbol: POLR2J2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032959.5 | NP_116581.3 | 4 | 468 | ||
GenBank mRNA | AB209398.1 | - | 3 | 1333 | ||
BC056864.1 | - | 4 | 358 | |||
BC086857.1 | - | 3 | 293 | 6007730 |
Gene Symbol: POLR2J3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097615.2 | NP_001091084.2 | 4 | 440 | ||
XM_017012372.1 | XP_016867861.1 | 4 | 479 | |||
XM_017012373.1 | XP_016867862.1 | 4 | 479 | |||
GenBank mRNA | AJ277740.1 | - | 3 | 165 | ||
AK309487.1 | - | 3 | 227 | |||
DA574442.1 | - | 3 | 228 | |||
DA580738.1 | - | 4 | 402 |
Gene Symbol: UPK3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030570.3 | NP_085047.1 | 5 | 1525 | ||
NM_182683.2 | NP_872624.1 | 6 | 1360 | |||
NM_182684.2 | NP_872625.1 | 7 | 1440 | |||
GenBank mRNA | AK056760.1 | - | 6 | 1168 | ||
AK290096.1 | - | 5 | 1428 | |||
AY233462.1 | - | 7 | 1273 | |||
BC004304.1 | - | 5 | 1349 | 3625247 |