Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112734.3 | NP_001106205.1 | 4 | 1078 | ||
NM_001166012.2 | NP_001159484.1 | 5 | 1282 | |||
NM_001288711.1 | NP_001275640.1 | 2 | 375 | |||
XM_005258183.4 | XP_005258240.1 | 4 | 1047 | |||
XM_006722371.3 | XP_006722434.1 | 5 | 1211 | |||
XM_011525789.2 | XP_011524091.1 | 1 | 276 | |||
XM_017025515.1 | XP_016881004.1 | 5 | 1230 | |||
XM_017025516.1 | XP_016881005.1 | 4 | 1063 | |||
XM_017025517.1 | XP_016881006.1 | 4 | 1057 | |||
XM_017025518.1 | XP_016881007.1 | 4 | 1129 | |||
XM_017025519.1 | XP_016881008.1 | 4 | 1034 | |||
XM_017025520.1 | XP_016881009.1 | 3 | 928 | |||
XM_017025521.1 | XP_016881010.1 | 3 | 881 | |||
XM_017025522.1 | XP_016881011.1 | 3 | 860 | |||
GenBank mRNA | AF303374.1 | - | 1 | 500 | ||
AK303447.1 | - | 5 | 1266 | |||
AY234408.1 | - | 4 | 1041 | |||
DA653861.1 | - | 2 | 360 |
Gene Symbol: ZNF397 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006722558.2 | XP_006722621.1 | 4 | 9626 | ||
XM_011526229.2 | XP_011524531.1 | 5 | 9715 | |||
XM_011526230.2 | XP_011524532.1 | 4 | 9917 | |||
XM_011526231.2 | XP_011524533.1 | 4 | 9514 | |||
XM_017026042.1 | XP_016881531.1 | 4 | 9569 |