Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136504.1 | NP_001129976.1 | 9 | 5926 | ||
NM_177402.4 | NP_796376.2 | 9 | 5916 | |||
XM_011509192.2 | XP_011507494.1 | 9 | 5946 | |||
XM_017000309.1 | XP_016855798.1 | 8 | 7692 | |||
XM_017000310.1 | XP_016855799.1 | 8 | 7683 | |||
XM_017000311.1 | XP_016855800.1 | 9 | 5935 | |||
XM_017000312.1 | XP_016855801.1 | 9 | 6600 | |||
XM_017000313.1 | XP_016855802.1 | 9 | 6590 | |||
GenBank mRNA | AK124510.1 | - | 1 | 2237 |