Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MALRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142308.2 | NP_001135780.2 | 34 | 5971 | ||
XM_011519453.2 | XP_011517755.1 | 35 | 6509 | |||
XM_011519454.1 | XP_011517756.1 | 35 | 5835 | |||
XM_011519455.2 | XP_011517757.1 | 34 | 6336 | |||
XM_017016182.1 | XP_016871671.1 | 34 | 5781 | |||
XM_017016183.1 | XP_016871672.1 | 35 | 6510 | |||
XM_017016184.1 | XP_016871673.1 | 23 | 4198 | |||
XM_017016185.1 | XP_016871674.1 | 23 | 4195 | |||
XM_017016186.1 | XP_016871675.1 | 16 | 2752 | |||
GenBank mRNA | AK297683.1 | - | 6 | 854 | ||
KC843478.1 | - | 34 | 5971 |