Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142531.1 | NP_001136003.1 | 6, 7 | 1332 | ||
NM_001142532.1 | NP_001136004.1 | 6, 7 | 1581 | |||
NM_001142533.1 | NP_001136005.1 | 5, 6 | 1469 | |||
NM_001142534.1 | NP_001136006.1 | 5, 6 | 1441 | |||
NM_001252334.1 | NP_001239263.1 | 7, 8 | 1838 | |||
NM_005489.3 | NP_005480.2 | 6, 7 | 1467 | |||
NM_170600.2 | NP_733745.1 | 7, 8 | 1940 | |||
XM_005251639.1 | XP_005251696.1 | 6, 7 | 1448 | |||
XM_011518114.1 | XP_011516416.1 | 7, 8 | 1924 | |||
XM_011518115.1 | XP_011516417.1 | 7, 8 | 1884 | |||
XM_011518117.2 | XP_011516419.1 | 5, 6 | 2560 | |||
XM_017014174.1 | XP_016869663.1 | 5, 6 | 1721 | |||
GenBank mRNA | AF124251.1 | - | 6, 7 | 1379 | ||
AK056068.1 | - | 5, 6 | 1469 | |||
AK096983.1 | - | 3, 4 | 762 | |||
AK128287.1 | - | 8, 9 | 1871 | |||
AK291393.1 | - | 5, 6 | 1433 | |||
AK292041.1 | - | 6, 7 | 1395 | |||
AK292718.1 | - | 7, 8 | 1811 | |||
AK294386.1 | - | 6, 7 | 1332 | |||
AY358089.1 | - | 6, 7 | 1449 | |||
AY358440.1 | - | 6, 7 | 1368 | |||
BC027962.1 | - | 6, 7 | 1338 | 5223012 | ||
BC032365.1 | - | 7, 8 | 1815 | 4521962 | ||
BX647905.1 | - | 7, 8 | 1764 | |||
DA054525.1 | - | 1, 2 | 532 |