Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSFM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172695.1 | NP_001166166.1 | 2 | 237 | ||
NM_001172696.1 | NP_001166167.1 | 2 | 237 | |||
NM_001172697.1 | NP_001166168.1 | 2 | 237 | |||
NM_005726.5 | NP_005717.3 | 2 | 237 | |||
GenBank mRNA | AF110399.1 | - | 2 | 183 | ||
AK295323.1 | - | 2 | 201 | |||
AK304621.1 | - | 2 | 221 | |||
AK308981.1 | - | 2 | 200 | |||
AK313453.1 | - | 2 | 221 | |||
BC022862.1 | - | 2 | 210 | 4076880 | ||
BC093068.1 | - | 2 | 192 | 30722527 | ||
DB448773.1 | - | 2 | 237 | |||
L37936.1 | - | 1 | 88 |