Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005729.3 | NP_005720.1 | 4 | 464 | ||
XM_005269379.2 | XP_005269436.1 | 4 | 462 | |||
GenBank mRNA | AK296669.1 | - | 4 | 457 | ||
AK312757.1 | - | 4 | 459 | |||
BC005020.1 | - | 4 | 450 | 3659599 | ||
BC013204.1 | - | 4 | 441 | |||
BC110299.1 | - | 4 | 441 | 5215228 | ||
BE791220.1 | - | 4 | 480 | |||
M80254.1 | - | 4 | 462 |