Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTDSP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005730.3 | NP_005721.3 | 8 | 1261 | ||
XM_005268556.1 | XP_005268613.1 | 8 | 1039 | |||
GenBank mRNA | AF000152.1 | - | 8 | 1037 | ||
AF022231.1 | - | 8 | 978 | |||
AK226076.1 | - | 8 | 1017 | |||
AK291289.1 | - | 8 | 1008 | |||
AK294930.1 | - | 3 | 408 | |||
AK310183.1 | - | 4 | 482 | |||
AK310873.1 | - | 8 | 763 | |||
AK311070.1 | - | 3 | 311 | |||
AK312413.1 | - | 8 | 1020 | |||
AY279531.1 | - | 8 | 732 | |||
BC018922.2 | - | 8 | 993 | |||
BC065920.1 | - | 8 | 976 | 5756196 |