Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037164.2 | NP_001032241.1 | 2 | 846 | ||
NM_001195396.1 | NP_001182325.1 | 2 | 1069 | |||
NM_005738.4 | NP_005729.1 | 2 | 1072 | |||
NM_212460.3 | NP_997625.1 | 2 | 843 | |||
GenBank mRNA | AF493890.1 | - | 1 | 540 | ||
AK313233.1 | - | 2 | 725 | |||
BC001111.2 | - | 2 | 762 | 3050172 | ||
BC003027.1 | - | 2 | 762 | 2905330 | ||
BM550177.1 | - | 2 | 692 | |||
CD049640.1 | - | 2 | 801 | |||
U73960.1 | - | 2 | 693 |
Gene Symbol: NSUN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017008615.1 | XP_016864104.1 | 8 | 1658 |