Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABI2 SIRNA Assays ›
Gene Symbol:
ABI2
Gene Name:
abl-interactor 2
Gene Aliases:
ABI-2, ABI2B, AIP-1, AblBP3, SSH3BP2, argBP1, argBPIA, argBPIB
Chromosome Location:
Chr.2: 203327874 - 203432173 on Build GRCh38
UniGene ID:
Hs.471156
Species:
Human
siRNA ID
HSS173425
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ABI2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282925.1
NP_001269854.1
10
1571
NM_001282926.1
NP_001269855.1
9
1553
NM_001282927.1
NP_001269856.1
8
1046
NM_001282932.1
NP_001269861.1
9
1567
NM_005759.5
NP_005750.4
9
1457
XM_005246217.2
XP_005246274.1
11
2023
XM_005246218.2
XP_005246275.1
10
2005
XM_005246221.2
XP_005246278.1
9
1858
XM_005246225.2
XP_005246282.1
8
1771
XM_005246226.2
XP_005246283.1
8
1735
XM_006712174.2
XP_006712237.1
11
2023
XM_006712177.2
XP_006712240.1
12
2099
XM_006712179.2
XP_006712242.1
10
1876
XM_006712180.2
XP_006712243.1
10
1840
XM_006712181.2
XP_006712244.1
9
1753
XM_006712182.2
XP_006712245.1
9
1750
XM_006712185.1
XP_006712248.1
9
1811
XM_006712186.2
XP_006712249.1
7
1588
XM_011510472.2
XP_011508774.1
10
1873
XM_011510473.2
XP_011508775.1
10
1322
XM_017003118.1
XP_016858607.1
11
2020
XM_017003119.1
XP_016858608.1
11
2017
XM_017003120.1
XP_016858609.1
11
2020
XM_017003121.1
XP_016858610.1
10
1933
XM_017003122.1
XP_016858611.1
10
1870
XM_017003123.1
XP_016858612.1
9
1855
XM_017003124.1
XP_016858613.1
9
1822
XM_017003125.1
XP_016858614.1
9
1819
XM_017003126.1
XP_016858615.1
9
1816
XM_017003127.1
XP_016858616.1
9
1819
XM_017003128.1
XP_016858617.1
11
2012
XM_017003129.1
XP_016858618.1
9
1789
XM_017003130.1
XP_016858619.1
9
1786
XM_017003131.1
XP_016858620.1
8
1768
XM_017003132.1
XP_016858621.1
9
1753
XM_017003133.1
XP_016858622.1
11
1952
XM_017003134.1
XP_016858623.1
8
1735
XM_017003135.1
XP_016858624.1
8
1732
XM_017003136.1
XP_016858625.1
8
1732
XM_017003137.1
XP_016858626.1
11
1913
XM_017003138.1
XP_016858627.1
9
1690
XM_017003139.1
XP_016858628.1
10
1898
XM_017003140.1
XP_016858629.1
10
1895
XM_017003141.1
XP_016858630.1
8
1675
XM_017003142.1
XP_016858631.1
8
1672
XM_017003143.1
XP_016858632.1
10
1862
XM_017003144.1
XP_016858633.1
8
1606
XM_017003145.1
XP_016858634.1
7
1585
XM_017003146.1
XP_016858635.1
7
1588
GenBank mRNA
AF260261.1
-
10
1295
AK294429.1
-
4
623
AK297537.1
-
5
512
AK299092.1
-
8
789
AK299672.1
-
9
948
AK299679.1
-
4
489
AK299688.1
-
6
749
AK299750.1
-
4
395
AK299824.1
-
9
1397
AK302853.1
-
9
1493
AK316343.1
-
4
449
BC001439.2
-
9
1206
3138341
BT009920.1
-
9
1181
U23435.1
-
8
1007
X95632.1
-
9
1281
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
movement of cell or subcellular component
cytoskeleton organization
actin polymerization or depolymerization
viral process
cell migration
Rac protein signal transduction
peptidyl-tyrosine phosphorylation
positive regulation of Arp2/3 complex-mediated actin nucleation
DNA binding
protein binding
cytoskeletal adaptor activity
SH3 domain binding
kinase binding
ubiquitin protein ligase binding
protein complex binding
Rac GTPase binding
proline-rich region binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0