Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALYREF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005782.3 | NP_005773.3 | 4, 5 | 617 | ||
GenBank mRNA | AF047002.1 | - | 4, 5 | 518 | ||
AF086513.1 | - | 3, 4 | 272 | |||
BC052302.1 | - | 4, 5 | 617 | 6381528 | ||
BU624466.1 | - | 2, 3 | 493 |