Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127708.2 | NP_001121180.2 | 8 | 3470 | ||
NM_001127709.2 | NP_001121181.2 | 7 | 3314 | |||
NM_001127710.2 | NP_001121182.2 | 6 | 3191 | |||
NM_001303232.1 | NP_001290161.1 | 8 | 3464 | |||
NM_005807.4 | NP_005798.3 | 9 | 3593 | |||
XM_017000002.1 | XP_016855491.1 | 6 | 3610 | |||
XM_017000003.1 | XP_016855492.1 | 5 | 3177 | |||
GenBank mRNA | AK057197.1 | - | 7 | 1429 | ||
AK131434.1 | - | 8 | 2164 | |||
U70136.1 | - | 8 | 3575 |