Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278784.1 | NP_001265713.1 | 5 | 606 | ||
NM_005827.2 | NP_005818.2 | 5 | 631 | |||
XM_005256929.3 | XP_005256986.1 | 6 | 546 | |||
XM_006721632.3 | XP_006721695.1 | 6 | 750 | |||
XM_011524179.2 | XP_011522481.1 | 4 | 516 | |||
XM_011524180.2 | XP_011522482.1 | 5 | 482 | |||
GenBank mRNA | AK098056.1 | - | 3 | 1706 | ||
AK293563.1 | - | 5 | 631 | |||
AK296073.1 | - | 5 | 600 | |||
BC011888.2 | - | 5 | 610 | 3832062 | ||
BM476321.1 | - | 1 | 41 | |||
BX641166.1 | - | 5 | 885 | |||
CR745911.1 | - | 3 | 215 | |||
CX164572.1 | - | 3 | 238 | |||
D87989.1 | - | 5 | 600 |