Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRVI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098579.2 | NP_001092049.2 | 16 | 2153 | ||
NM_001100163.2 | NP_001093633.1 | 17 | 2431 | |||
NM_001100167.2 | NP_001093637.1 | 17 | 2148 | |||
NM_001206880.1 | NP_001193809.1 | 16 | 1866 | |||
NM_001206881.1 | NP_001193810.1 | 15 | 2273 | |||
NM_130385.3 | NP_569056.4 | 17 | 2484 | |||
GenBank mRNA | AF081249.1 | - | 15 | 2169 | ||
AF081250.1 | - | 15 | 1897 | |||
AK127209.1 | - | 17 | 2126 | |||
AK296336.1 | - | 17 | 2248 | |||
AK300358.1 | - | 16 | 2138 | |||
AK301296.1 | - | 15 | 1427 | |||
AK304702.1 | - | 16 | 1500 | |||
BC117127.1 | - | 17 | 2021 | 40125678 |