Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199573.1 | NP_001186502.1 | 4 | 921 | ||
NM_006074.4 | NP_006065.2 | 4 | 933 | |||
GenBank mRNA | AK290434.1 | - | 4 | 663 | ||
AK298715.1 | - | 5 | 613 | |||
AK298934.1 | - | 4 | 487 | |||
AK308796.1 | - | 3 | 553 | |||
AK310098.1 | - | 4 | 620 | |||
AL360134.1 | - | 2 | 91 | |||
BC022281.1 | - | 4 | 740 | 4718024 | ||
BC035582.1 | - | 4 | 706 | 5583800 | ||
HQ842635.1 | - | 3 | 554 | |||
X82200.1 | - | 4 | 664 |