Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMG20B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006339.2 | NP_006330.2 | 3 | 150 | ||
XM_017026144.1 | XP_016881633.1 | 3 | 157 | |||
GenBank mRNA | AF146223.1 | - | 2 | 92 | ||
AF288679.1 | - | 2 | 82 | |||
AF318366.1 | - | 2 | 391 | |||
AF331191.1 | - | 2 | 82 | |||
AK090733.1 | - | 2 | 116 | |||
AK289500.1 | - | 3 | 138 | |||
AK310601.1 | - | 2 | 116 | |||
AK314201.1 | - | 3 | 124 | |||
AL355691.1 | - | 1 | 48 | |||
AL355702.1 | - | 1 | 47 | |||
AL355703.1 | - | 2 | 94 | |||
AL355709.1 | - | 2 | 92 | |||
BC002552.2 | - | 3 | 140 | 3138919 | ||
BC003505.1 | - | 3 | 138 | 3609369 | ||
BC004408.1 | - | 2 | 88 | 3638942 | ||
BC021585.1 | - | 1 | 52 | 4873122 | ||
BI561304.1 | - | 3 | 163 |