Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PDLIM5 SIRNA Assays ›
Gene Symbol:
PDLIM5
Gene Name:
PDZ and LIM domain 5
Gene Aliases:
ENH, ENH1, L9, LIM
Chromosome Location:
Chr.4: 94451857 - 94668227 on Build GRCh38
UniGene ID:
Hs.480311
Species:
Human
siRNA ID
HSS173778
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PDLIM5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001011513.3
NP_001011513.3
6
699
NM_001011515.2
NP_001011515.1
7
717
NM_001011516.2
NP_001011516.1
8
657
NM_001256425.1
NP_001243354.1
5
607
NM_001256426.1
NP_001243355.1
7
717
NM_001256427.1
NP_001243356.1
7
717
NM_001256428.1
NP_001243357.1
5
874
NM_006457.4
NP_006448.4
6
1026
XM_005262693.4
XP_005262750.1
7
1014
XM_005262695.4
XP_005262752.1
6
996
XM_005262696.4
XP_005262753.1
6
614
XM_005262698.4
XP_005262755.1
7
639
XM_006714066.3
XP_006714129.1
8
1029
XM_006714068.3
XP_006714131.1
8
647
XM_006714069.3
XP_006714132.1
8
1029
XM_006714070.3
XP_006714133.1
8
630
XM_011531543.2
XP_011529845.1
7
612
XM_017007657.1
XP_016863146.1
7
1014
XM_017007658.1
XP_016863147.1
8
647
GenBank mRNA
AF061258.1
-
6
928
AI201986.1
-
3
355
AK291624.1
-
6
948
AK291898.1
-
8
582
AK296980.1
-
5
788
AL832157.1
-
6
601
AL833438.1
-
7
602
AY598328.1
-
5
845
BC008741.2
-
6
965
3345715
BC026029.2
-
4
469
BT007328.1
-
5
845
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of synapse assembly
regulation of dendritic spine morphogenesis
cell growth involved in cardiac muscle cell development
cell-cell adhesion
actin binding
protein kinase C binding
protein binding
zinc ion binding
actinin binding
protein N-terminus binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0