Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGF2BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007225.1 | NP_001007226.1 | 8 | 990 | ||
NM_001291869.1 | NP_001278798.1 | 8 | 1008 | |||
NM_001291872.1 | NP_001278801.1 | 7 | 810 | |||
NM_001291873.1 | NP_001278802.1 | 7 | 792 | |||
NM_001291874.1 | NP_001278803.1 | 7 | 792 | |||
NM_001291875.1 | NP_001278804.1 | 6 | 728 | |||
NM_006548.4 | NP_006539.3 | 8 | 990 | |||
XM_006713477.3 | XP_006713540.1 | 3 | 414 | |||
XM_006713478.3 | XP_006713541.1 | 3 | 409 | |||
XM_017005557.1 | XP_016861046.1 | 8 | 987 | |||
XM_017005558.1 | XP_016861047.1 | 8 | 1013 | |||
XM_017005559.1 | XP_016861048.1 | 8 | 1014 | |||
XM_017005560.1 | XP_016861049.1 | 8 | 910 | |||
XM_017005561.1 | XP_016861050.1 | 8 | 1015 | |||
XM_017005562.1 | XP_016861051.1 | 8 | 1014 | |||
XM_017005563.1 | XP_016861052.1 | 8 | 1014 | |||
XM_017005564.1 | XP_016861053.1 | 8 | 1014 | |||
GenBank mRNA | AF117107.1 | - | 8 | 957 | ||
AK290664.1 | - | 8 | 991 | |||
AK296703.1 | - | 3 | 254 | |||
AK309611.1 | - | 8 | 944 | |||
BC021290.2 | - | 8 | 975 | 5090334 | ||
EU408701.1 | - | 7 | 810 | |||
EU408702.1 | - | 7 | 792 | |||
EU408703.1 | - | 7 | 792 | |||
EU408704.1 | - | 6 | 728 |