Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CORIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278585.1 | NP_001265514.1 | 15 | 2143 | ||
NM_006587.3 | NP_006578.2 | 17 | 2455 | |||
GenBank mRNA | AF113248.1 | - | 1 | 100 | ||
AF133845.1 | - | 17 | 2391 | |||
AK304038.1 | - | 16 | 2220 | |||
BC110451.2 | - | 17 | 2391 | 40027293 |