Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CETN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297765.1 | NP_001284694.1 | 2 | 191 | ||
NM_001297768.1 | NP_001284697.1 | 2 | 191 | |||
NM_004365.3 | NP_004356.2 | 2 | 191 | |||
GenBank mRNA | BC005383.1 | - | 2 | 161 | 3935674 | |
BC093793.1 | - | 2 | 127 | 7939638 | ||
BC112040.1 | - | 2 | 127 | 8327508 | ||
BM553126.1 | - | 2 | 171 | |||
BT006688.1 | - | 2 | 66 | |||
BU569574.1 | - | 3 | 140 | |||
CB988525.1 | - | 2 | 213 | |||
CB998764.1 | - | 2 | 194 | |||
CR450346.1 | - | 2 | 66 | |||
Y12473.1 | - | 3 | 66 |