Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006613.3 | NP_006604.1 | 4, 5 | 557 | ||
XM_017024029.1 | XP_016879518.1 | 5, 6 | 689 | |||
GenBank mRNA | BC035856.1 | - | 4, 5 | 478 | 5768298 | |
BC063035.1 | - | 4, 5 | 529 | 5757176 | ||
BQ056077.1 | - | 4, 5 | 557 | |||
BT019981.1 | - | 4, 5 | 460 | |||
CR541941.1 | - | 4, 5 | 460 | |||
CR541966.1 | - | 4, 5 | 460 | |||
U52518.1 | - | 4, 5 | 493 |