Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDCCAG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039707.1 | NP_001034796.1 | 7 | 1117 | ||
NM_001039708.1 | NP_001034797.1 | 5 | 898 | |||
NM_006643.3 | NP_006634.3 | 6 | 1048 | |||
XM_005266050.3 | XP_005266107.1 | 7 | 1071 | |||
XM_005266051.3 | XP_005266108.1 | 6 | 962 | |||
XM_011518157.2 | XP_011516459.1 | 6 | 901 | |||
XM_017014218.1 | XP_016869707.1 | 6 | 1218 | |||
XM_017014219.1 | XP_016869708.1 | 6 | 1218 | |||
GenBank mRNA | AF039688.1 | - | 3 | 185 | ||
AK001296.1 | - | 5 | 677 | |||
AY129014.1 | - | 1 | 594 | |||
BC014515.1 | - | 6 | 915 | 3547130 |