Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBGCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256617.1 | NP_001243546.1 | 16, 17 | 2754 | ||
NM_001256618.1 | NP_001243547.1 | 14, 15 | 2481 | |||
NM_006659.3 | NP_006650.1 | 15, 16 | 2670 | |||
NR_046330.1 | 15, 16 | 3017 | ||||
XM_017015595.1 | XP_016871084.1 | 8, 9 | 1139 | |||
XM_017015596.1 | XP_016871085.1 | 8, 9 | 1139 | |||
GenBank mRNA | AF042379.1 | - | 15, 16 | 2360 | ||
AK092113.1 | - | 15, 16 | 3017 | |||
AK096087.1 | - | 7, 8 | 2148 | |||
AK223586.1 | - | 15, 16 | 2561 | |||
AK297251.1 | - | 14, 15 | 2203 | |||
AK299564.1 | - | 15, 16 | 2224 | |||
AK314520.1 | - | 14, 15 | 2461 | |||
BC093770.1 | - | 14, 15 | 2349 | 7939615 | ||
BC111957.1 | - | 14, 15 | 2349 | 8327425 | ||
BC143247.1 | - | 15, 16 | 2433 | 9051749 |