Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYVE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006691.3 | NP_006682.2 | 6 | 1169 | ||
GenBank mRNA | AF118108.1 | - | 6 | 973 | ||
AF127670.2 | - | 6 | 1003 | |||
AK075443.1 | - | 6 | 1083 | |||
AK300885.1 | - | 4 | 616 | |||
AK312462.1 | - | 6 | 1167 | |||
AY358925.1 | - | 6 | 1042 | |||
BC026231.1 | - | 6 | 928 | 4712863 |