Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300861.1 | NP_001287790.1 | 7 | 881 | ||
NM_020470.2 | NP_065203.2 | 8 | 1037 | |||
XM_005273720.3 | XP_005273777.1 | 7 | 1102 | |||
XM_017017139.1 | XP_016872628.1 | 7 | 1138 | |||
XM_017017140.1 | XP_016872629.1 | 8 | 1049 | |||
GenBank mRNA | AF004876.1 | - | 8 | 970 | ||
AK226003.1 | - | 8 | 1015 | |||
AK291124.1 | - | 8 | 1015 | |||
BC001299.1 | - | 8 | 988 | 3451489 | ||
BC009892.2 | - | 5 | 464 | |||
CR456710.1 | - | 8 | 855 |