Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006713.3 | NP_006704.3 | 3 | 265 | ||
XM_011513944.2 | XP_011512246.1 | 4 | 518 | |||
XM_017008986.1 | XP_016864475.1 | 4 | 361 | |||
XM_017008987.1 | XP_016864476.1 | 4 | 238 | |||
GenBank mRNA | AB209285.1 | - | 3 | 160 | ||
AK294124.1 | - | 3 | 207 | |||
AK307268.1 | - | 3 | 197 | |||
AK311754.1 | - | 3 | 207 | |||
BC009610.1 | - | 3 | 193 | 4297490 | ||
BC010537.1 | - | 3 | 168 | 3457167 | ||
BC018189.1 | - | 3 | 187 | 3881373 | ||
BC022339.1 | - | 3 | 198 | 4134117 | ||
BC063402.1 | - | 3 | 181 | |||
BC071655.1 | - | 3 | 182 | |||
BC105972.1 | - | 3 | 163 | 3907729 | ||
BX472106.1 | - | 3 | 266 | |||
CR456902.1 | - | 2 | 137 | |||
U12979.1 | - | 2 | 137 | |||
X79805.1 | - | 2 | 137 |