Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOMM34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006809.4 | NP_006800.2 | 4 | 613 | ||
XM_005260254.2 | XP_005260311.1 | 4 | 546 | |||
XM_011528501.1 | XP_011526803.1 | 4 | 432 | |||
XM_017027600.1 | XP_016883089.1 | 4 | 660 | |||
GenBank mRNA | AB085681.1 | - | 4 | 538 | ||
AK000594.1 | - | 4 | 532 | |||
AK222952.1 | - | 4 | 520 | |||
AK302128.1 | - | 4 | 442 | |||
AK307655.1 | - | 4 | 554 | |||
BC001763.1 | - | 4 | 526 | 3544124 | ||
BC007423.2 | - | 4 | 526 | 2960090 | ||
BC014907.1 | - | 4 | 503 | 3922502 | ||
BI560064.1 | - | 4 | 632 | |||
BT020008.1 | - | 4 | 473 | |||
BT020009.1 | - | 4 | 473 | |||
CR456765.1 | - | 4 | 473 |