Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPRE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143826.2 | NP_001137298.1 | 5 | 588 | ||
NM_001143827.2 | NP_001137299.1 | 5 | 734 | |||
NM_001256420.1 | NP_001243349.1 | 3 | 548 | |||
NM_014268.3 | NP_055083.1 | 4 | 676 | |||
NR_046177.1 | 4 | 491 | ||||
GenBank mRNA | AB016823.1 | - | 4 | 611 | ||
AK090945.1 | - | 3 | 504 | |||
AK294833.1 | - | 5 | 678 | |||
AK296251.1 | - | 4 | 491 | |||
AK315766.1 | - | 4 | 629 | |||
BC007318.2 | - | 4 | 619 | 2989643 | ||
BT020086.1 | - | 4 | 496 | |||
CR536545.1 | - | 4 | 496 | |||
X94232.1 | - | 4 | 636 |