Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LILRB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081442.2 | NP_001074911.1 | 6 | 1332 | ||
NM_001081443.2 | NP_001074912.1 | 5 | 1032 | |||
NM_001304457.1 | NP_001291386.1 | 6 | 1305 | |||
NM_006840.4 | NP_006831.1 | 6 | 1332 | |||
XM_011526359.2 | XP_011524661.1 | 6 | 1316 | |||
XM_011526360.2 | XP_011524662.1 | 6 | 1316 | |||
XM_011526361.2 | XP_011524663.1 | 6 | 1316 | |||
XM_011526362.2 | XP_011524664.1 | 5 | 997 | |||
GenBank mRNA | AF025534.1 | - | 6 | 1283 | ||
AK057072.1 | - | 5 | 986 | |||
AK090399.1 | - | 3 | 1319 | |||
AK092867.1 | - | 6 | 1200 | |||
AK096122.1 | - | 4 | 359 | |||
AK223296.1 | - | 6 | 1288 | |||
AK290845.1 | - | 6 | 1316 | |||
BC025704.1 | - | 6 | 1286 | 5223581 |