Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LILRB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081442.2 | NP_001074911.1 | 5 | 868 | ||
NM_001081443.2 | NP_001074912.1 | 4 | 568 | |||
NM_001304457.1 | NP_001291386.1 | 5 | 841 | |||
NM_006840.4 | NP_006831.1 | 5 | 868 | |||
XM_011526359.2 | XP_011524661.1 | 5 | 852 | |||
XM_011526360.2 | XP_011524662.1 | 5 | 852 | |||
XM_011526361.2 | XP_011524663.1 | 5 | 852 | |||
XM_011526362.2 | XP_011524664.1 | 4 | 533 | |||
GenBank mRNA | AF025534.1 | - | 5 | 819 | ||
AK057072.1 | - | 4 | 522 | |||
AK090399.1 | - | 2 | 855 | |||
AK092867.1 | - | 5 | 736 | |||
AK290845.1 | - | 5 | 852 | |||
AK301112.1 | - | 5 | 804 | |||
AK301338.1 | - | 5 | 601 | |||
BC025704.1 | - | 5 | 822 | 5223581 |