Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMILIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007046.3 | NP_008977.1 | 8 | 3492 | ||
GenBank mRNA | AF088916.1 | - | 8 | 2993 | ||
AL050138.1 | - | 8 | 2923 | |||
BC007530.1 | - | 9 | 1950 | |||
BC009947.2 | - | 5 | 1687 | |||
BC136279.1 | - | 8 | 3092 | 9020266 | ||
BC142638.1 | - | 8 | 2220 | |||
BC142688.1 | - | 8 | 3374 | 40148654 |