Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006703.3 | NP_006694.1 | 4 | 603 | ||
GenBank mRNA | AF062529.1 | - | 4 | 438 | ||
AF062530.1 | - | 4 | 438 | |||
AK313440.1 | - | 4 | 506 | |||
BC007727.2 | - | 4 | 564 | 4303483 | ||
BG714472.1 | - | 4 | 602 | |||
BT019984.1 | - | 4 | 285 | |||
BT019985.1 | - | 4 | 285 |
Gene Symbol: RPS10-NUDT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202470.2 | NP_001189399.1 | 8 | 736 | ||
GenBank mRNA | BU521288.1 | - | 8 | 659 |