Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102575.1 | NP_001096045.1 | 1 | 1559 | ||
NM_001145427.1 | NP_001138899.1 | 1 | 1563 | |||
NM_052870.2 | NP_443102.2 | 1 | 1559 | |||
XM_017008997.1 | XP_016864486.1 | 1 | 1563 | |||
GenBank mRNA | AF395536.1 | - | 1 | 1369 | ||
AK304195.1 | - | 1 | 1563 | |||
BC060791.1 | - | 1 | 1559 | 30341956 | ||
BC067860.1 | - | 1 | 1559 | 30343220 | ||
BC117218.1 | - | 1 | 1457 | 40125769 | ||
BC117220.1 | - | 1 | 1457 | 40125771 |