Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007249.4 | NP_009180.3 | 3 | 272 | ||
XM_005266251.3 | XP_005266308.1 | 3 | 381 | |||
XM_011534907.2 | XP_011533209.1 | 2 | 95 | |||
XM_011534908.2 | XP_011533210.1 | 2 | 95 | |||
XM_011534909.2 | XP_011533211.1 | 3 | 228 | |||
XM_011534910.2 | XP_011533212.1 | 2 | 59 | |||
XM_011534911.2 | XP_011533213.1 | 2 | 137 | |||
XM_011534912.2 | XP_011533214.1 | 2 | 208 | |||
XM_017020384.1 | XP_016875873.1 | 2 | 39 | |||
XM_017020385.1 | XP_016875874.1 | 2 | 138 | |||
GenBank mRNA | AF113122.1 | - | 3 | 246 | ||
AF161471.1 | - | 3 | 120 | |||
AF330041.1 | - | 2 | 58 | |||
AJ243274.1 | - | 2 | 50 | |||
AK291397.1 | - | 3 | 368 | |||
BC019680.1 | - | 2 | 77 | 4942405 | ||
DB125574.1 | - | 3 | 117 | |||
HF546212.1 | - | 3 | 243 |