Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007263.3 | NP_009194.2 | 8 | 800 | ||
NM_199442.1 | NP_955474.1 | 7 | 647 | |||
NM_199444.1 | NP_955476.1 | 7 | 644 | |||
XM_011527655.1 | XP_011525957.1 | 9 | 943 | |||
XM_011527656.1 | XP_011525958.1 | 8 | 787 | |||
GenBank mRNA | AI680336.1 | - | 3 | 338 | ||
AJ131182.1 | - | 8 | 800 | |||
AK222584.1 | - | 8 | 797 | |||
AK315281.1 | - | 8 | 797 | |||
CR456886.1 | - | 8 | 758 |