Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145047.1 | NP_001138519.1 | 10, 11 | 748 | ||
NM_001145048.1 | NP_001138520.1 | 9, 10 | 680 | |||
NM_001145049.1 | NP_001138521.1 | 7, 8 | 573 | |||
NM_007266.3 | NP_009197.2 | 9, 10 | 787 | |||
NR_026735.1 | 7, 8 | 680 | ||||
GenBank mRNA | AB044661.1 | - | 9, 10 | 748 | ||
AJ010842.1 | - | 9, 10 | 677 | |||
AK298827.1 | - | 9, 10 | 1081 | |||
AK298866.1 | - | 9, 10 | 787 | |||
AK302131.1 | - | 7, 8 | 646 | |||
AK310340.1 | - | 10, 11 | 738 | |||
AK311423.1 | - | 7, 8 | 703 | |||
AK312992.1 | - | 9, 10 | 759 | |||
AK316404.1 | - | 7, 8 | 573 | |||
BC007451.2 | - | 9, 10 | 731 | 3029376 |