Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK11IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052902.3 | NP_443134.3 | 19 | 2481 | ||
XM_011510526.1 | XP_011508828.1 | 19 | 2461 | |||
XM_011510527.2 | XP_011508829.1 | 19 | 2461 | |||
XM_011510528.2 | XP_011508830.1 | 8 | 1408 | |||
GenBank mRNA | AB067485.1 | - | 17 | 2159 | ||
AF450267.1 | - | 19 | 2435 | |||
BC014114.2 | - | 10 | 1522 | |||
BC034051.1 | - | 19 | 2458 | 4819148 |