Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145798.2 | NP_665741.1 | 5 | 537 | ||
XM_017024122.1 | XP_016879611.1 | 5 | 559 | |||
XM_017024123.1 | XP_016879612.1 | 5 | 677 | |||
XM_017024124.1 | XP_016879613.1 | 4 | 524 | |||
GenBank mRNA | AF323729.1 | - | 5 | 526 | ||
AF392446.1 | - | 5 | 590 | |||
AK289523.1 | - | 5 | 559 | |||
BC028010.1 | - | 5 | 536 | |||
BC065482.1 | - | 5 | 509 | 6081784 |